Assay Details
Target Gene Details
Entrez Gene ID: | 123624 |
Gene Name: | ATP/GTP binding protein like 1 |
Gene Aliases: |
CCP4, FECD8 |
Location: |
Chr.15:86079635-87031476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGBL1 | XM_011521226.2 | XP_011519528.1 | ||
XM_011521227.2 | XP_011519529.1 | |||
XM_011521228.2 | XP_011519530.1 | |||
XM_011521230.2 | XP_011519532.1 | |||
XM_017021918.1 | XP_016877407.1 | |||
XM_017021919.1 | XP_016877408.1 | |||
XM_017021920.1 | XP_016877409.1 | |||
XM_017021921.1 | XP_016877410.1 |
Target Gene Details
Entrez Gene ID: | 101929701 |
Gene Name: | long intergenic non-protein coding RNA 1584 |
Gene Aliases: |
- |
Location: |
Chr.15:86083345-86116717 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01584 | NR_120368.1 | |||
BX116292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1648 | Chr.15:86078294 - 86119577 on Build GRCh38 | Deletion | LINC01584 AGBL1 |
nsv519797 | Chr.15:86084875 - 87238541 on Build GRCh38 | Gain | LINC01584 AGBL1 LOC105370954 AGBL1-AS1 LOC102724452 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |