Assay Details
Target Gene Details
Entrez Gene ID: | 390598 |
Gene Name: | SKI family transcriptional corepressor 1 |
Gene Aliases: |
CORL1, FUSSEL15, LBXCOR1 |
Location: |
Chr.15:67819704-67834561 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SKOR1 | NM_001258024.1 | NP_001244953.1 | ||
AY669507.1 | AAV74188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv827375 | Chr.15:67819001 - 67830434 on Build GRCh38 | Gain | SKOR1 |
nsv1041463 | Chr.15:67815528 - 67849977 on Build GRCh38 | Gain | SKOR1 LOC101929076 RNU6-1 |
nsv569803 | Chr.15:67585107 - 68124237 on Build GRCh38 | Gain | PIAS1 MAP2K5 SKOR1 LOC101929076 RNU6-1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |