Assay Details
Target Gene Details
Entrez Gene ID: | 10021 |
Gene Name: | hyperpolarization activated cyclic nucleotide gated potassium channel 4 |
Gene Aliases: |
SSS2 |
Location: |
Chr.15:73319859-73369264 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HCN4 | NM_005477.2 | NP_005468.1 | ||
AJ132429.1 | CAB42604.1 | |||
AJ238850.1 | CAB52754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524049 | Chr.15:73320492 - 73387960 on Build GRCh38 | Loss |
![]() ![]() |
nsv569949 | Chr.15:73320036 - 73387960 on Build GRCh38 | Loss |
![]() ![]() |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)