Assay Details
Target Gene Details
Entrez Gene ID: | 771 |
Gene Name: | carbonic anhydrase 12 |
Gene Aliases: |
CA-XII, CAXII, HsT18816, T18816 |
Location: |
Chr.15:63321378-63382166 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CA12 | NM_001218.4 | NP_001209.1 | ||
NM_001293642.1 | NP_001280571.1 | |||
NM_206925.2 | NP_996808.1 | |||
NR_135511.1 | ||||
AF037335.1 | AAC63952.1 | |||
AF051882.1 | AAC39789.1 | |||
AK096845.1 | ||||
AK315769.1 | ||||
BC000278.1 | AAH00278.1 | |||
BC011691.2 | AAH11691.1 | |||
BC023981.1 | AAH23981.1 | |||
BT006656.1 | AAP35302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)