Assay Details
Target Gene Details
Entrez Gene ID: | 10240 |
Gene Name: | mitochondrial ribosomal protein S31 |
Gene Aliases: |
IMOGN38, MRP-S31, S31mt |
Location: |
Chr.13:40729296-40771220 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRPS31 | NM_005830.3 | NP_005821.2 | ||
AK315248.1 | ||||
BC022045.1 | AAH22045.1 | |||
Z68747.1 | CAA92951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892332 | Chr.13:40619338 - 40820761 on Build GRCh38 | Gain | MIR320D1 MIR621 MRPS31 TPTE2P5 SLC25A15 FOXO1 |
nsv974239 | Chr.13:40724480 - 40774465 on Build GRCh38 | Duplication | MIR320D1 MRPS31 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |