Assay Details
Target Gene Details
Entrez Gene ID: | 10150 |
Gene Name: | muscleblind like splicing regulator 2 |
Gene Aliases: |
MBLL, MBLL39, PRO2032 |
Location: |
Chr.13:97141780-97394120 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBNL2 | XM_011521045.2 | XP_011519347.1 | ||
XM_017020308.1 | XP_016875797.1 | |||
XM_017020309.1 | XP_016875798.1 | |||
XM_017020310.1 | XP_016875799.1 | |||
XM_017020311.1 | XP_016875800.1 | |||
CR749802.1 | CAH18662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1137 | Chr.13:97151220 - 97196560 on Build GRCh38 | Deletion | MBNL2 LINC00456 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |