Assay Details
Target Gene Details
Entrez Gene ID: | 420 |
Gene Name: | ADP-ribosyltransferase 4 (Dombrock blood group) |
Gene Aliases: |
ARTC4, CD297, DO, DOK1 |
Location: |
Chr.12:14829311-14843479 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ART4 | NM_021071.2 | NP_066549.2 | ||
AF290204.1 | AAG17845.1 | |||
AK291662.1 | ||||
AY690666.1 | AAW02794.1 | |||
AY829431.1 | AAX39378.1 | |||
AY829432.1 | AAX39379.1 | |||
AY829434.1 | AAX39381.1 | |||
BC074727.2 | AAH74727.1 | |||
BX096478.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)