Assay Details
Target Gene Details
Entrez Gene ID: | 266743 |
Gene Name: | neuronal PAS domain protein 4 |
Gene Aliases: |
Le-PAS, NXF, PASD10, bHLHe79 |
Location: |
Chr.11:66409158-66426707 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPAS4 | XM_017017537.1 | XP_016873026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832193 | Chr.11:66352868 - 66511334 on Build GRCh38 | Loss | LOC101928069 SLC29A2 PELI3 DPP3 NPAS4 MRPL11 LOC102724064 BBS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |