Assay Details
Target Gene Details
Entrez Gene ID: | 196264 |
Gene Name: | myelin protein zero like 3 |
Gene Aliases: |
- |
Location: |
Chr.11:118226690-118252368 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MPZL3 | NM_001286152.1 | NP_001273081.1 | ||
NM_198275.2 | NP_938016.1 | |||
NR_104404.1 | ||||
NR_104405.1 | ||||
AK095399.1 | ||||
AK289390.1 | ||||
AK296951.1 | ||||
AK304292.1 | ||||
AK310966.1 | ||||
AY358626.1 | AAQ88989.1 | |||
BC113586.1 | ||||
BC143897.1 | ||||
DC417020.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |