Assay Details
Target Gene Details
Entrez Gene ID: | 116071 |
Gene Name: | basic leucine zipper ATF-like transcription factor 2 |
Gene Aliases: |
SARI |
Location: |
Chr.11:64987945-64997045 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BATF2 | NM_138456.3 | NP_612465.3 | ||
AK096702.1 | BAC04846.1 | |||
HM134154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2750762 | Chr.11:64991485 - 64993636 on Build GRCh38 | Loss | BATF2 |
nsv555208 | Chr.11:64877647 - 65003707 on Build GRCh38 | Loss | MIR194-2HG GPHA2 PPP2R5B EHD1 MIR192 MAJIN ATG2A BATF2 MIR6749 MIR6750 MIR194-2 |
nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain | TIGD3 MIR612 MIR6879 SLC25A45 ATG2A SNX15 LOC101927789 ARL2-SNX15 CDCA5 ZNHIT2 MIR194-2 MIR6751 CDC42EP2 VPS51 FRMD8 POLA2 FAU BATF2 TM7SF2 CAPN1 MIR6749 MIR6750 GPHA2 PPP2R5B DPF2 NEAT1 MRPL49 TMEM262 ZFPL1 ARL2 MIR194-2HG SLC22A20 EHD1 MIR192 SAC3D1 NAALADL1 SYVN1 MAJIN SPDYC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |