Assay Details
Target Gene Details
Entrez Gene ID: | 55293 |
Gene Name: | UEV and lactate/malate dehyrogenase domains |
Gene Aliases: |
ATTP, UEV3 |
Location: |
Chr.11:18529609-18588837 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UEVLD | NM_001040697.2 | NP_001035787.1 | ||
NM_001261382.1 | NP_001248311.1 | |||
NM_001261383.1 | NP_001248312.1 | |||
NM_001261384.1 | NP_001248313.1 | |||
NM_001261385.1 | NP_001248314.1 | |||
NM_001261386.1 | NP_001248315.1 | |||
NM_018314.4 | NP_060784.3 | |||
XM_006718255.2 | XP_006718318.1 | |||
XM_011520201.2 | XP_011518503.1 | |||
XM_017017985.1 | XP_016873474.1 | |||
XM_017017986.1 | XP_016873475.1 | |||
AF503350.1 | AAN32950.1 | |||
AK001930.1 | BAA91985.1 | |||
AK295501.1 | ||||
AK296835.1 | ||||
AK301537.1 | ||||
AK314521.1 | ||||
AK316191.1 | ||||
BC064566.1 | AAH64566.1 | |||
BU633352.1 | ||||
CD109744.1 | ||||
CF138618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain | SPTY2D1-AS1 TSG101 LDHA TMEM86A MIR3159 MRGPRX1 LDHC IGSF22 GTF2H1 UEVLD HPS5 SPTY2D1 PTPN5 LDHAL6A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |