Assay Details
Target Gene Details
Entrez Gene ID: | 26297 |
Gene Name: | secretion regulating guanine nucleotide exchange factor |
Gene Aliases: |
DELGEF, Gnefr |
Location: |
Chr.11:17788048-18013162 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERGEF | NM_012139.3 | NP_036271.1 | ||
NR_104040.1 | ||||
NR_104041.1 | ||||
AF086056.1 | ||||
AJ243950.1 | CAB60832.1 | |||
AJ243951.1 | CAB60833.1 | |||
AK292286.1 | ||||
BC065375.1 | AAH65375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain | NCR3LG1 OTOG OR7E14P LOC105376575 RPS13 LOC102723330 KCNJ11 PLEKHA7 PIK3C2A NUCB2 MYOD1 KCNC1 USH1C SNORD14A SERGEF ABCC8 SNORD14B |
nsv509390 | Chr.11:17726193 - 17855296 on Build GRCh38 | Insertion | KCNC1 SERGEF |
nsv832078 | Chr.11:17758956 - 17931974 on Build GRCh38 | Loss | KCNC1 SERGEF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |