Assay Details
Target Gene Details
Entrez Gene ID: | 8495 |
Gene Name: | PPFIA binding protein 2 |
Gene Aliases: |
Cclp1 |
Location: |
Chr.11:7513765-7667329 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPFIBP2 | NM_003621.3 | NP_003612.2 | ||
XM_005253177.2 | XP_005253234.1 | |||
XM_006718349.2 | XP_006718412.1 | |||
XM_006718350.2 | XP_006718413.1 | |||
XM_011520413.2 | XP_011518715.1 | |||
XM_011520415.2 | XP_011518717.1 | |||
XM_011520416.2 | XP_011518718.1 | |||
XM_017018443.1 | XP_016873932.1 | |||
AK001131.1 | ||||
AK308398.1 | ||||
BC021714.1 | AAH21714.1 | |||
BG720018.1 | ||||
DA339729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625293 | Chr.11:7515685 - 7685956 on Build GRCh38 | Gain | PPFIBP2 CYB5R2 OVCH2 |
esv3625294 | Chr.11:7523852 - 7629549 on Build GRCh38 | Gain | PPFIBP2 |
nsv7659 | Chr.11:7512472 - 7557142 on Build GRCh38 | Deletion | PPFIBP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |