Assay Details
Target Gene Details
Entrez Gene ID: | 64421 |
Gene Name: | DNA cross-link repair 1C |
Gene Aliases: |
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C |
Location: |
Chr.10:14897853-14954432 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469720 | Chr.10:14865281 - 15037379 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv68n27 | Chr.10:14913179 - 15018884 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv961n54 | Chr.10:14910793 - 15028616 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv831793 | Chr.10:14870604 - 15058763 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)