Assay Details
Target Gene Details
Entrez Gene ID: | 657 |
Gene Name: | bone morphogenetic protein receptor type 1A |
Gene Aliases: |
10q23del, ACVRLK3, ALK3, CD292, SKR5 |
Location: |
Chr.10:86755786-86927969 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BMPR1A | NM_004329.2 | NP_004320.2 | ||
XM_011540103.2 | XP_011538405.1 | |||
XM_011540104.2 | XP_011538406.1 | |||
AB451405.1 | ||||
AK291764.1 | ||||
BC028383.1 | AAH28383.1 | |||
Z22535.1 | CAA80257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss | MINPP1 FAM25A NUTM2A-AS1 LINC00863 NUTM2A LINC00864 GLUD1 NUTM2D AGAP11 BMPR1A MMRN2 SNCG MIR4678 FAM35A ADIRF |
esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss | MINPP1 FAM25A NUTM2A-AS1 LINC00863 NUTM2A LINC00864 GLUD1 NUTM2D AGAP11 BMPR1A MMRN2 SNCG MIR4678 FAM35A ADIRF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |