Assay Details
Target Gene Details
Entrez Gene ID: | 2849 |
Gene Name: | G protein-coupled receptor 26 |
Gene Aliases: |
- |
Location: |
Chr.10:123666355-123697399 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR26 | NM_153442.3 | NP_703143.1 | ||
AJ505757.1 | CAD44281.1 | |||
BC112011.1 | AAI12012.1 | |||
BC113462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv467469 | Chr.10:123668548 - 123688189 on Build GRCh38 | Loss | GPR26 |
nsv832011 | Chr.10:123590498 - 123760281 on Build GRCh38 | Gain+Loss | CPXM2 GPR26 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |