Assay Details
Target Gene Details
Entrez Gene ID: | 5264 |
Gene Name: | phytanoyl-CoA 2-hydroxylase |
Gene Aliases: |
LN1, LNAP1, PAHX, PHYH1, RD |
Location: |
Chr.10:13277796-13300130 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHYH | NM_001037537.1 | NP_001032626.1 | ||
NM_001323080.1 | NP_001310009.1 | |||
NM_001323082.1 | NP_001310011.1 | |||
NM_001323083.1 | NP_001310012.1 | |||
NM_001323084.1 | NP_001310013.1 | |||
NM_006214.3 | NP_006205.1 | |||
AF023462.1 | AAB81834.1 | |||
AF112977.1 | AAD20602.1 | |||
BC029512.1 | AAH29512.1 | |||
CR542055.1 | CAG46852.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)