Assay Details
Target Gene Details
Entrez Gene ID: | 338599 |
Gene Name: | dual specificity phosphatase and pro isomerase domain containing 1 |
Gene Aliases: |
DUSP27, FMDSP |
Location: |
Chr.10:75026427-75073827 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DUPD1 | NM_001003892.1 | NP_001003892.1 | ||
XM_011539747.2 | XP_011538049.1 | |||
AY686755.1 | AAT94288.1 | |||
BC137321.1 | ||||
BC137322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv926n100 | Chr.10:74994872 - 75081593 on Build GRCh38 | Gain |
![]() ![]() |
dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)