Assay Details
Target Gene Details
Entrez Gene ID: | 57512 |
Gene Name: | G protein-coupled receptor 158 |
Gene Aliases: |
- |
Location: |
Chr.10:25174722-25602229 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR158 | NM_020752.2 | NP_065803.2 | ||
AK095120.1 | ||||
AY528411.1 | AAS18315.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |