Assay Details
Target Gene Details
Entrez Gene ID: | 2869 |
Gene Name: | G protein-coupled receptor kinase 5 |
Gene Aliases: |
GPRK5 |
Location: |
Chr.10:119207685-119455619 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRK5 | NM_005308.2 | NP_005299.1 | ||
XM_005269707.1 | XP_005269764.1 | |||
XM_005269708.1 | XP_005269765.1 | |||
AK298261.1 | ||||
AK309761.1 | ||||
AK313012.1 | ||||
BC064506.1 | AAH64506.1 | |||
L15388.1 | AAA58620.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)