Assay Details
Target Gene Details
Entrez Gene ID: | 65217 |
Gene Name: | protocadherin related 15 |
Gene Aliases: |
CDHR15, DFNB23, USH1F |
Location: |
Chr.10:53802771-55629182 on Build GRCh38 |
Assay Gene Location: | Within Intron 44 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCDH15 | NM_001142769.1 | NP_001136241.1 | ||
NM_001142770.1 | NP_001136242.1 | |||
NM_001142771.1 | NP_001136243.1 | |||
NM_001142772.1 | NP_001136244.1 | |||
XM_017016571.1 | XP_016872060.1 | |||
XM_017016572.1 | XP_016872061.1 | |||
XM_017016573.1 | XP_016872062.1 | |||
EU718480.1 | ||||
EU718481.1 | ||||
EU718482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041917 | Chr.10:52741572 - 55682853 on Build GRCh38 | Loss | LOC105378311 MIR548F1 MBL2 PCDH15 MTRNR2L5 |
esv2750962 | Chr.10:52972795 - 55555375 on Build GRCh38 | Loss | LOC105378311 MIR548F1 PCDH15 |
dgv861n100 | Chr.10:53792127 - 53821891 on Build GRCh38 | Loss | PCDH15 |
esv2759753 | Chr.10:53597376 - 53827170 on Build GRCh38 | Loss | PCDH15 |
dgv1146n54 | Chr.10:53034448 - 54727838 on Build GRCh38 | Loss | LOC105378311 MIR548F1 PCDH15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |