Assay Details
Target Gene Details
Entrez Gene ID: | 142910 |
Gene Name: | lipase family member J |
Gene Aliases: |
LIPL1, bA425M17.2 |
Location: |
Chr.10:88583914-88623161 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPJ | XM_017015740.1 | XP_016871229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551839 | Chr.10:88575078 - 88662379 on Build GRCh38 | Loss | LIPJ RNLS |
esv3624134 | Chr.10:88555882 - 88703649 on Build GRCh38 | Gain | LIPF LIPJ RNLS |
esv3624131 | Chr.10:88489037 - 88688695 on Build GRCh38 | Loss | LIPF LIPJ RNLS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |