Assay Details
Target Gene Details
Entrez Gene ID: | 83938 |
Gene Name: | chromosome 10 open reading frame 11 |
Gene Aliases: |
CDA017 |
Location: |
Chr.10:75431646-76559220 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C10orf11 | NM_001305581.1 | NP_001292510.1 | ||
XM_011540256.2 | XP_011538558.1 | |||
XM_011540257.2 | XP_011538559.1 | |||
XM_017016759.1 | XP_016872248.1 | |||
XM_017016760.1 | XP_016872249.1 | |||
XM_017016761.1 | XP_016872250.1 | |||
BM720401.1 | ||||
BX105111.1 | ||||
HY006640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv825472 | Chr.10:75435954 - 75441201 on Build GRCh38 | Loss |
![]() |
esv3623851 | Chr.10:75436041 - 75441393 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)