Assay Details
Target Gene Details
Entrez Gene ID: | 338567 |
Gene Name: | potassium two pore domain channel subfamily K member 18 |
Gene Aliases: |
K2p18.1, MGR13, TRESK, TRESK-2, TRESK2, TRIK |
Location: |
Chr.10:117197489-117210299 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNK18 | NM_181840.1 | NP_862823.1 | ||
AB087138.1 | BAC78527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3624674 | Chr.10:117141857 - 117425204 on Build GRCh38 | Gain | PDZD8 MIR3663HG KCNK18 SLC18A2 MIR3663 |
nsv832002 | Chr.10:117037559 - 117246329 on Build GRCh38 | Gain+Loss | MIR3663HG VAX1 SHTN1 KCNK18 SLC18A2 MIR3663 |
esv34075 | Chr.10:117199852 - 117208093 on Build GRCh38 | Loss | KCNK18 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |