Assay Details
Target Gene Details
Entrez Gene ID: | 84159 |
Gene Name: | AT-rich interaction domain 5B |
Gene Aliases: |
DESRT, MRF-2, MRF2 |
Location: |
Chr.10:61901254-62096948 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARID5B | NM_032199.2 | NP_115575.1 | ||
XM_011540262.1 | XP_011538564.1 | |||
AK296921.1 | ||||
BC107800.1 | AAI07801.1 | |||
BG615173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv8681 | Chr.10:61896798 - 62671529 on Build GRCh38 | Gain | LOC283045 RTKN2 ARID5B ZNF365 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |