Assay Details
Target Gene Details
Entrez Gene ID: | 26103 |
Gene Name: | leucine rich repeat, Ig-like and transmembrane domains 1 |
Gene Aliases: |
FIGLER9, LRRC21, PAL |
Location: |
Chr.10:84231520-84241461 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRIT1 | NM_015613.2 | NP_056428.1 | ||
AB031547.1 | BAA95681.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1046749 | Chr.10:83270389 - 84629720 on Build GRCh38 | Gain | CCSER2 GHITM LINC00858 C10orf99 LOC105378397 HOST2 CDHR1 LRIT2 LRIT1 RGR |
nsv551745 | Chr.10:83260366 - 84632635 on Build GRCh38 | Gain | CCSER2 GHITM LINC00858 C10orf99 LOC105378397 HOST2 CDHR1 LRIT2 LRIT1 RGR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |