Assay Details
Target Gene Details
Entrez Gene ID: | 100188947 |
Gene Name: | HECTD2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:91306962-91611460 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECTD2-AS1 | NR_024467.1 | |||
AA211589.1 | ||||
BQ182529.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)