Assay Details
Target Gene Details
Entrez Gene ID: | 159195 |
Gene Name: | ubiquitin specific peptidase 54 |
Gene Aliases: |
C10orf29, bA137L10.3, bA137L10.4 |
Location: |
Chr.10:73497538-73626844 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)