Assay Details
Target Gene Details
Entrez Gene ID: | 79723 |
Gene Name: | suppressor of variegation 3-9 homolog 2 |
Gene Aliases: |
KMT1B |
Location: |
Chr.10:14878783-14904315 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUV39H2 | NM_001193424.1 | NP_001180353.1 | ||
NM_001193425.1 | NP_001180354.1 | |||
NM_001193426.1 | NP_001180355.1 | |||
NM_001193427.1 | NP_001180356.1 | |||
NM_024670.3 | NP_078946.1 | |||
NR_034181.1 | ||||
XM_006717503.3 | XP_006717566.1 | |||
XM_011519662.2 | XP_011517964.1 | |||
XM_017016637.1 | XP_016872126.1 | |||
AK027067.1 | BAB15645.1 | |||
AK222565.1 | ||||
AK225369.1 | ||||
BC007754.2 | AAH07754.1 | |||
BC029360.1 | AAH29360.1 | |||
BU928392.1 | ||||
DB040298.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469720 | Chr.10:14865281 - 15037379 on Build GRCh38 | Gain | LOC105376430 SUV39H2 MEIG1 HSPA14 DCLRE1C LOC105376432 |
nsv831793 | Chr.10:14870604 - 15058763 on Build GRCh38 | Loss | LOC105376430 SUV39H2 MEIG1 HSPA14 OLAH DCLRE1C LOC105376432 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |