Assay Details
Target Gene Details
Entrez Gene ID: | 6934 |
Gene Name: | transcription factor 7 like 2 |
Gene Aliases: |
TCF-4, TCF4 |
Location: |
Chr.10:112950219-113167678 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762881 | Chr.10:112985316 - 112991059 on Build GRCh38 | Loss | TCF7L2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |