Assay Details
Target Gene Details
Entrez Gene ID: | 55753 |
Gene Name: | oxoglutarate dehydrogenase-like |
Gene Aliases: |
- |
Location: |
Chr.10:49734641-49762379 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OGDHL | NM_001143996.1 | NP_001137468.1 | ||
NM_001143997.1 | NP_001137469.1 | |||
NM_018245.2 | NP_060715.2 | |||
XM_011539946.1 | XP_011538248.1 | |||
XM_011539947.1 | XP_011538249.1 | |||
XM_017016401.1 | XP_016871890.1 | |||
XM_017016402.1 | XP_016871891.1 | |||
XM_017016403.1 | XP_016871892.1 | |||
AB033116.3 | ||||
AK001713.1 | BAA91855.1 | |||
AK225558.1 | ||||
AK290226.1 | ||||
AK296551.1 | ||||
AK303729.1 | ||||
AK307759.1 | ||||
BC026320.1 | AAH26320.1 | |||
DA411851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048905 | Chr.10:49720681 - 49811144 on Build GRCh38 | Gain |
![]() |
nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)