Assay Details
Target Gene Details
Entrez Gene ID: | 5654 |
Gene Name: | HtrA serine peptidase 1 |
Gene Aliases: |
ARMD7, CADASIL2, CARASIL, HtrA, L56, ORF480, PRSS11 |
Location: |
Chr.10:122461525-122514908 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTRA1 | NM_002775.4 | NP_002766.1 | ||
AF097709.1 | AAC97211.1 | |||
AK091944.1 | ||||
AK092476.1 | ||||
AK290089.1 | ||||
BC011352.1 | ||||
D87258.1 | BAA13322.1 | |||
DA633502.1 | ||||
Y07921.1 | CAA69226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv7580 | Chr.10:122446276 - 122490980 on Build GRCh38 | Deletion |
![]() ![]() |
nsv825594 | Chr.10:122434264 - 122505709 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)