Assay Details
Target Gene Details
Entrez Gene ID: | 256536 |
Gene Name: | transcription elongation regulator 1 like |
Gene Aliases: |
- |
Location: |
Chr.10:131092391-131311721 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TCERG1L | NM_174937.3 | NP_777597.2 | ||
XM_011539577.1 | XP_011537879.1 | |||
AK096269.1 | ||||
BC042951.1 | AAH42951.1 | |||
BC093639.1 | AAH93639.1 | |||
BC101535.1 | AAI01536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832027 | Chr.10:131056320 - 131214052 on Build GRCh38 | Gain | TCERG1L TCERG1L-AS1 |
nsv1042634 | Chr.10:131116646 - 131247825 on Build GRCh38 | Loss | TCERG1L |
nsv1048253 | Chr.10:129847356 - 131188858 on Build GRCh38 | Gain | TCERG1L MIR378C GLRX3 LOC102724883 EBF3 LINC00959 CTAGE7P TCERG1L-AS1 |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | TCERG1L ADGRA1-AS1 BNIP3 LOC105378568 CFAP46 DPYSL4 LINC01167 NKX6-2 LINC01168 PPP2R2D TCERG1L-AS1 LINC01164 INPP5A PWWP2B LOC107984282 MIR378C LRRC27 ADGRA1 STK32C LINC01166 JAKMIP3 C10orf91 |
nsv428242 | Chr.10:130607951 - 131583141 on Build GRCh38 | Gain | TCERG1L MIR378C TCERG1L-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |