Assay Details
Target Gene Details
Entrez Gene ID: | 51703 |
Gene Name: | acyl-CoA synthetase long-chain family member 5 |
Gene Aliases: |
ACS2, ACS5, FACL5 |
Location: |
Chr.10:112374158-112428380 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACSL5 | NM_016234.3 | NP_057318.2 | ||
NM_203379.1 | NP_976313.1 | |||
NM_203380.1 | NP_976314.1 | |||
XM_017016324.1 | XP_016871813.1 | |||
AB033899.1 | BAA85979.1 | |||
AK000339.1 | ||||
AK024932.1 | ||||
AK222782.1 | ||||
AK223288.1 | ||||
AL708480.1 | ||||
AY358520.1 | AAQ88884.1 | |||
BC007985.2 | AAH07985.2 | |||
CD689921.1 | ||||
CF595490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508609 | Chr.10:112338315 - 112385553 on Build GRCh38 | Deletion | ACSL5 GUCY2GP |
nsv7552 | Chr.10:112340440 - 112383097 on Build GRCh38 | Deletion | ACSL5 GUCY2GP |
nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss | ACSL5 VTI1A ZDHHC6 GUCY2GP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |