Assay Details
Target Gene Details
Entrez Gene ID: | 57584 |
Gene Name: | Rho GTPase activating protein 21 |
Gene Aliases: |
ARHGAP10 |
Location: |
Chr.10:24583609-24725421 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP21 | NM_020824.3 | NP_065875.3 | ||
XM_005252542.3 | XP_005252599.1 | |||
XM_005252544.4 | XP_005252601.1 | |||
XM_011519602.2 | XP_011517904.1 | |||
XM_011519603.2 | XP_011517905.1 | |||
XM_011519604.2 | XP_011517906.1 | |||
XM_011519605.2 | XP_011517907.1 | |||
XM_011519606.2 | XP_011517908.1 | |||
XM_011519607.2 | XP_011517909.1 | |||
XM_017016459.1 | XP_016871948.1 | |||
XM_017016460.1 | XP_016871949.1 | |||
XM_017016461.1 | XP_016871950.1 | |||
AB037845.2 | BAA92662.2 | |||
AF480466.1 | AAM22955.1 | |||
AK090642.1 | ||||
AK295649.1 | ||||
AK297278.1 | ||||
AK307604.1 | ||||
AL834495.1 | CAD39153.1 | |||
BC118915.2 | ||||
BX537570.1 | CAD97787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6099 | Chr.10:24597662 - 24627801 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)