Assay Details
Target Gene Details
Entrez Gene ID: | 8027 |
Gene Name: | signal transducing adaptor molecule |
Gene Aliases: |
STAM-1, STAM1 |
Location: |
Chr.10:17644125-17716824 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STAM | NM_001324282.1 | NP_001311211.1 | ||
NM_001324283.1 | NP_001311212.1 | |||
NM_001324284.1 | NP_001311213.1 | |||
NM_001324285.1 | NP_001311214.1 | |||
NM_001324286.1 | NP_001311215.1 | |||
NM_001324287.1 | NP_001311216.1 | |||
NM_001324288.1 | NP_001311217.1 | |||
NM_001324289.1 | NP_001311218.1 | |||
NM_003473.3 | NP_003464.1 | |||
XM_011519695.2 | XP_011517997.1 | |||
AK303083.1 | ||||
AK314405.1 | ||||
AU118708.1 | ||||
BC030586.2 | AAH30586.1 | |||
U43899.1 | AAC50734.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)