Assay Details
Target Gene Details
Entrez Gene ID: | 8829 |
Gene Name: | neuropilin 1 |
Gene Aliases: |
BDCA4, CD304, NP1, NRP, VEGF165R |
Location: |
Chr.10:33177491-33334905 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRP1 | NM_001244972.1 | NP_001231901.1 | ||
NM_001244973.1 | NP_001231902.1 | |||
NM_003873.5 | NP_003864.4 | |||
NR_045259.1 | ||||
XM_006717521.1 | XP_006717584.1 | |||
XM_006717522.1 | XP_006717585.1 | |||
XM_006717523.2 | XP_006717586.1 | |||
XM_006717524.1 | XP_006717587.1 | |||
XM_006717525.1 | XP_006717588.1 | |||
XM_017016865.1 | XP_016872354.1 | |||
XM_017016866.1 | XP_016872355.1 | |||
AB209641.1 | BAD92878.1 | |||
AF016050.1 | AAC12921.1 | |||
AF018956.1 | AAC51759.1 | |||
AK292822.1 | ||||
AL833608.1 | CAH10397.1 | |||
AY249243.1 | AAP80144.1 | |||
BX510902.1 | CAD91133.1 | |||
BX648250.1 | ||||
CR749333.1 | CAH18187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831829 | Chr.10:33178304 - 33344113 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)