Assay Details
Target Gene Details
Entrez Gene ID: | 221037 |
Gene Name: | jumonji domain containing 1C |
Gene Aliases: |
TRIP-8, TRIP8 |
Location: |
Chr.10:63167221-63522075 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JMJD1C | NM_001282948.1 | NP_001269877.1 | ||
NM_001318153.1 | NP_001305082.1 | |||
NM_001318154.1 | NP_001305083.1 | |||
NM_001322252.1 | NP_001309181.1 | |||
NM_001322254.1 | NP_001309183.1 | |||
NM_001322258.1 | NP_001309187.1 | |||
NM_032776.2 | NP_116165.1 | |||
NR_134512.1 | ||||
XM_011539503.1 | XP_011537805.1 | |||
XM_017015897.1 | XP_016871386.1 | |||
XM_017015898.1 | XP_016871387.1 | |||
XM_017015899.1 | XP_016871388.1 | |||
XM_017015900.1 | XP_016871389.1 | |||
XM_017015901.1 | XP_016871390.1 | |||
XM_017015902.1 | XP_016871391.1 | |||
XM_017015903.1 | XP_016871392.1 | |||
AB037801.1 | BAA92618.1 | |||
AL831917.1 | CAD38578.1 | |||
BC143722.1 | ||||
BX537954.1 | CAD97921.1 | |||
EF068222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2677353 | Chr.10:63130283 - 63501631 on Build GRCh38 | Deletion | MIR1296 LOC105378330 JMJD1C-AS1 NRBF2 JMJD1C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |