Assay Details
Target Gene Details
Entrez Gene ID: | 221037 |
Gene Name: | jumonji domain containing 1C |
Gene Aliases: |
TRIP-8, TRIP8 |
Location: |
Chr.10:63167221-63522075 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2677353 | Chr.10:63130283 - 63501631 on Build GRCh38 | Deletion | JMJD1C MIR1296 JMJD1C-AS1 LOC105378330 NRBF2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |