Assay Details
Target Gene Details
Entrez Gene ID: | 3416 |
Gene Name: | insulin degrading enzyme |
Gene Aliases: |
INSULYSIN |
Location: |
Chr.10:92451684-92574484 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IDE | NM_001165946.1 | NP_001159418.1 | ||
NM_001322793.1 | NP_001309722.1 | |||
NM_001322794.1 | NP_001309723.1 | |||
NM_001322795.1 | NP_001309724.1 | |||
NM_001322796.1 | NP_001309725.1 | |||
NM_001322797.1 | NP_001309726.1 | |||
NM_004969.3 | NP_004960.2 | |||
NR_136399.1 | ||||
XM_017016187.1 | XP_016871676.1 | |||
XM_017016188.1 | XP_016871677.1 | |||
XM_017016189.1 | XP_016871678.1 | |||
XM_017016190.1 | XP_016871679.1 | |||
AB209204.1 | BAD92441.1 | |||
AK093287.1 | ||||
AK302578.1 | ||||
AK312810.1 | ||||
AK316407.1 | ||||
BC096336.1 | AAH96336.1 | |||
BC096337.1 | AAH96337.1 | |||
BC096339.3 | AAH96339.1 | |||
BX648462.1 | ||||
M21188.1 | AAA52712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508600 | Chr.10:92355268 - 92472160 on Build GRCh38 | Deletion |
![]() ![]() |
dgv1349n54 | Chr.10:92452985 - 92530727 on Build GRCh38 | Loss |
![]() |
nsv1046258 | Chr.10:92107212 - 92706349 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)