Assay Details
Target Gene Details
Entrez Gene ID: | 54764 |
Gene Name: | zinc finger RANBP2-type containing 1 |
Gene Aliases: |
TRABID |
Location: |
Chr.10:124916896-124988059 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZRANB1 | NM_017580.2 | NP_060050.2 | ||
XM_005269925.4 | XP_005269982.1 | |||
XM_005269926.4 | XP_005269983.1 | |||
XM_006717907.2 | XP_006717970.1 | |||
XM_017016357.1 | XP_016871846.1 | |||
XM_017016358.1 | XP_016871847.1 | |||
AJ252060.1 | CAB64449.1 | |||
AK302810.1 | ||||
AK307386.1 | ||||
AL832925.1 | CAH10620.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv552289 | Chr.10:124875780 - 124998761 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)