Assay Details
Target Gene Details
Entrez Gene ID: | 64106 |
Gene Name: | neuropeptide FF receptor 1 |
Gene Aliases: |
GPR147, NPFF1, NPFF1R1, OT7T022 |
Location: |
Chr.10:70254957-70283694 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPFFR1 | NM_022146.4 | NP_071429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss | TBATA TSPAN15 TYSND1 PALD1 H2AFY2 NPFFR1 PPA1 EIF4EBP2 AIFM2 ADAMTS14 COL13A1 NEUROG3 NODAL PRF1 LRRC20 C10orf35 SAR1A |
nsv1041698 | Chr.10:70244698 - 70306282 on Build GRCh38 | Gain | LRRC20 NPFFR1 |
nsv831908 | Chr.10:70210726 - 70389945 on Build GRCh38 | Gain | LRRC20 NPFFR1 PPA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |