Assay Details
Target Gene Details
Entrez Gene ID: | 54886 |
Gene Name: | phospholipid phosphatase related 1 |
Gene Aliases: |
LPPR1, PRG-3 |
Location: |
Chr.9:101028749-101325135 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLPPR1 | NM_207299.1 | NP_997182.1 | ||
AK303807.1 | ||||
AY304515.1 | AAP72152.1 | |||
BC022465.1 | AAH22465.1 | |||
BI601921.1 | ||||
BI914341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831669 | Chr.9:100946843 - 101115840 on Build GRCh38 | Gain | PLPPR1 |
esv3621244 | Chr.9:101050753 - 101074973 on Build GRCh38 | Loss | PLPPR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |