Assay Details
Target Gene Details
Entrez Gene ID: | 1902 |
Gene Name: | lysophosphatidic acid receptor 1 |
Gene Aliases: |
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1 |
Location: |
Chr.9:110873252-111039259 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527204 | Chr.9:110883984 - 110888767 on Build GRCh38 | Loss | LPAR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |