Assay Details
Target Gene Details
Entrez Gene ID: | 9442 |
Gene Name: | mediator complex subunit 27 |
Gene Aliases: |
CRAP34, CRSP34, CRSP8, MED3, TRAP37 |
Location: |
Chr.9:131860110-132079887 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED27 | NM_001253881.1 | NP_001240810.1 | ||
NM_004269.3 | NP_004260.2 | |||
XM_005272236.3 | XP_005272293.1 | |||
XM_017015329.1 | XP_016870818.1 | |||
XM_017015330.1 | XP_016870819.1 | |||
AF104252.1 | AAD12721.1 | |||
AF230382.1 | AAF37290.1 | |||
BC002878.1 | AAH02878.1 | |||
BC064608.1 | AAH64608.1 | |||
DQ099387.1 | AAZ13763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3621898 | Chr.9:131733919 - 131887845 on Build GRCh38 | Gain |
![]() ![]() |
nsv1038424 | Chr.9:131788536 - 131906562 on Build GRCh38 | Gain |
![]() |
esv3621896 | Chr.9:131651740 - 131887259 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)