Assay Details
Target Gene Details
Entrez Gene ID: | 115426 |
Gene Name: | ubiquitin like with PHD and ring finger domains 2 |
Gene Aliases: |
NIRF, RNF107, TDRD23, URF2 |
Location: |
Chr.9:6413148-6508108 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UHRF2 | NM_152896.2 | NP_690856.1 | ||
NR_046386.1 | ||||
XM_011517704.2 | XP_011516006.1 | |||
XM_017014253.1 | XP_016869742.1 | |||
XM_017014254.1 | XP_016869743.1 | |||
AB071698.1 | BAB68317.1 | |||
AF274049.1 | AAM33799.1 | |||
AK308433.1 | ||||
BC028397.2 | AAH28397.1 | |||
BG709183.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831497 | Chr.9:6377501 - 6612402 on Build GRCh38 | Gain | GLDC UHRF2 |
nsv1029665 | Chr.9:5687332 - 6775107 on Build GRCh38 | Gain | KDM4C MIR4665 KIAA2026 RIC1 GLDC TPD52L3 UHRF2 RANBP6 ERMP1 MLANA IL33 |
dgv7386n100 | Chr.9:6188652 - 6687205 on Build GRCh38 | Gain | GLDC TPD52L3 UHRF2 IL33 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |