Assay Details
Target Gene Details
Entrez Gene ID: | 2961 |
Gene Name: | general transcription factor IIE subunit 2 |
Gene Aliases: |
FE, TF2E2, TFIIE-B, TTD6 |
Location: |
Chr.8:30578514-30658241 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF2E2 | NM_002095.4 | NP_002086.1 | ||
XM_011544509.2 | XP_011542811.1 | |||
XM_011544510.1 | XP_011542812.1 | |||
XM_017013363.1 | XP_016868852.1 | |||
XM_017013364.1 | XP_016868853.1 | |||
BC030572.2 | AAH30572.1 | |||
S67861.1 | AAB20414.1 | |||
X63469.1 | CAA45069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526509 | Chr.8:30328056 - 30643198 on Build GRCh38 | Gain | RBPMS RBPMS-AS1 SMIM18 GTF2E2 |
esv3616790 | Chr.8:30614869 - 30623976 on Build GRCh38 | Loss | GTF2E2 |
esv2661538 | Chr.8:30614907 - 30623972 on Build GRCh38 | Deletion | GTF2E2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |