Assay Details
Target Gene Details
Entrez Gene ID: | 55284 |
Gene Name: | ubiquitin conjugating enzyme E2 W (putative) |
Gene Aliases: |
UBC-16, UBC16 |
Location: |
Chr.8:73780097-73878910 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2W | NM_001271015.1 | NP_001257944.1 | ||
BC010900.1 | AAH10900.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77488476] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |