Assay Details
Target Gene Details
Entrez Gene ID: | 203102 |
Gene Name: | ADAM metallopeptidase domain 32 |
Gene Aliases: |
- |
Location: |
Chr.8:39107529-39284917 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAM32 | NM_001313994.1 | NP_001300923.1 | ||
NM_145004.6 | NP_659441.3 | |||
XM_006716298.2 | XP_006716361.1 | |||
XM_006716299.1 | XP_006716362.1 | |||
XM_011544433.1 | XP_011542735.1 | |||
XM_011544434.1 | XP_011542736.1 | |||
XM_017013183.1 | XP_016868672.1 | |||
XM_017013184.1 | XP_016868673.1 | |||
XM_017013186.1 | XP_016868675.1 | |||
AK122608.1 | BAC85489.1 | |||
AK300148.1 | ||||
AK313864.1 | ||||
AY172952.1 | AAO45696.1 | |||
AY358739.1 | AAQ89099.1 | |||
BC026085.2 | AAH26085.1 | |||
BC026169.2 | AAH26169.1 | |||
BC028702.2 | AAH28702.1 | |||
BC030014.2 | AAH30014.1 | |||
BC030698.2 | AAH30698.1 | |||
BC034975.2 | AAH34975.1 | |||
BI559590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6162 | Chr.8:39101789 - 39134363 on Build GRCh38 | Insertion | ADAM32 ADAM9 |
More Information
Additional Information:
For this assay, SNP(s) [rs115687239] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |