Assay Details
Target Gene Details
Entrez Gene ID: | 54928 |
Gene Name: | inositol monophosphatase domain containing 1 |
Gene Aliases: |
GPAPP, IMP 3, IMP-3, IMPA3 |
Location: |
Chr.8:56957929-56993874 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IMPAD1 | NM_017813.4 | NP_060283.3 | ||
AK000428.1 | BAA91158.1 | |||
AY032885.1 | AAK52336.1 | |||
BC017797.1 | AAH17797.1 | |||
BC067814.1 | AAH67814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891400 | Chr.8:56933474 - 57202184 on Build GRCh38 | Gain | IMPAD1 |
nsv1029443 | Chr.8:56835555 - 57145097 on Build GRCh38 | Loss | IMPAD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |